Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD9 Antibody (002), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290291MFV610
Description
COMMD9 Monoclonal antibody specifically detects COMMD9 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
COMMD9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
29099 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
002 | |
Immunohistochemistry-Paraffin | |
COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166 | |
Rabbit immunized with purified, recombinant Human COMMD9 (Uniprot#: Q9P000-1, Met1-Lys198) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction