Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD9 Antibody (04), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306601APC
Description
COMMD9 Monoclonal antibody specifically detects COMMD9 in Human samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| COMMD9 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 29099 | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| 04 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COMMD9 (Uniprot#: Q9P000-1; Met1-Lys198). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction