Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD9 Antibody (04), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306601DL594
Description
COMMD9 Monoclonal antibody specifically detects COMMD9 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
COMMD9 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
29099 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
04 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COMMD9 (Q9P000-1; Met1-Lys198). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction