Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common beta Chain Antibody (OTI4H3), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270463AF700
Description
Common beta Chain Monoclonal specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Common beta Chain | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CSF2RB | |
Full length recombinant protein of human Common beta Chain (NP_000386) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4H3 | |
Western Blot, Immunohistochemistry | |
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
Mouse | |
Affinity Purified | |
Phospho Specific | |
1439.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction