Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Antibody (3E12), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP312039R
Description
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow CytometrySpecifications
Common gamma Chain/IL-2 R gamma | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
3000000000000 | |
Flow Cytometry | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt). | |
0.1 mL | |
Cancer | |
3561 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction