Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Antibody (MM0195-7A23) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Common gamma Chain/IL-2 R gamma |
---|---|
Clone | MM0195-7A23 |
Concentration | LYOPH |
Applications | Blocking Assay, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
Common gamma Chain/IL-2 R gamma | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
IL2RG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0195-7A23 | |
Blocking Assay, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer | |
P31785 | |
3561 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title