Learn More
Ampco Common Knife
Click to view available options
Length (English) Blade:
3 -1/8 in.
5 -3/4 in.
Description
- Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
- Conforms to ASTM F2503.
- Meets the requirements established by ASTM A342.
Specifications
Specifications
Product Type | Common Knife |
Length (English) Blade | 5 -3/4 in. |
Length (English) | 10 -1/4 in. |
Quantity | 1 Ea. |