Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMP Monoclonal Antibody (4H6G10D2)
GREENER_CHOICE

Catalog No. PIMA530266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530266 Supplier Invitrogen™ Supplier No. MA530266
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human COMP/THBS5.

COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMP
Applications ELISA
Classification Monoclonal
Clone 4H6G10D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene COMP
Gene Accession No. P49747
Gene Alias Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5
Gene Symbols COMP
Host Species Mouse
Immunogen Recombinant Human COMP protein (Met1-Ala757).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1311
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.