Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMP Polyclonal Antibody
COMP Polyclonal Antibody
Supplier: Thermo Scientific PA596435
Description
COMP Polyclonal Antibody for Western Blot, ICC/IF

Specifications
COMP | |
Polyclonal | |
Unconjugated | |
COMP | |
Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12845, 1311, 25304 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.36 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P35444, P49747, Q9R0G6 | |
COMP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-160 of human COMP (NP_0000862). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction