Learn More
Contec Multi Bucket System with Flat Mops
Shop All Contec Inc. Products
Click to view available options
Material:
Polypropylene Bucket, Stainless Steel Cart
Stainless Steel
Description
The Contec Multi-Bucket System is designed for use with Contec's flat mops, offering flexibility, mobility, and easy customization for cleanroom cleaning.
- Configurable with two or three buckets in stainless steel or polypropylene
- Extendable front frame accommodates an additional bucket for added capacity
- Quiet, locking casters ensure smooth movement and stability
- Utility handle for easy transport of cleaning supplies
Hands-free mop removal tool available for VertiKlean™ MAX™ and QuickTask™ mop heads
- Integrated mop holder with brace for secure storage during transport
- Compatible with stainless steel downpress wringer for flat mops
- Autoclavable at 121°C for 30 minutes for sterilization in critical environments
Specifications
Specifications
| Product Type | 2 Bucket System |
| Material | Polypropylene Bucket, Stainless Steel Cart |