Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3 (Active) Monoclonal Antibody (bH6)
GREENER_CHOICE

Catalog No. FERMA548991
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548991 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548991 Supplier Invitrogen™ Supplier No. MA548991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody bH6 is specific for a C3 neo-epitope expressed on the cleavage fragments of C3b, iC3b, and C3c, but not C3dg and C3f.

Complement C3 is a protein that plays a vital role in the immune system. It is a central component of the complement system, a group of proteins that work together to defend the body against infections. Complement C3 is produced by the liver and circulates in the blood. When an infection or injury occurs, complement C3 is activated and triggers a cascade of events that help to eliminate the invading pathogen or damaged tissue. However, abnormalities in complement C3 can also lead to autoimmune diseases and other health problems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3 (Active)
Applications Inhibition Assays, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone bH6
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene C3
Gene Accession No. P01024
Gene Alias Active C3; Active Complement C3; Complement C3
Gene Symbols C3
Host Species Mouse
Immunogen The monoclonal antibody bH6 recognizes a neo-epitope expressed on the cleavage fragments of C3b, iC3b, and C3c.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.