Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3 Antibody (F1-8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246609H
Description
Complement C3 Monoclonal specifically detects Complement C3 in Human samples. It is validated for ELISA.Specifications
Complement C3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
C3 | |
This Complement C3 Antibody (F1-8) was developed against Complement component 3 (C3) purified from human plasma. | |
0.1 mL | |
Primary | |
Antibody is specific for human complement component 3 (C3/C3b/C3c). | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA | |
F1-8 | |
ELISA | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Mouse | |
Protein A or G purified | |
Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
718 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction