Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3 Mouse anti-Human, Clone: F1-8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2466090.025MG
Description
Complement C3 Monoclonal specifically detects Complement C3 in Human samples. It is validated for ELISA, Immunoprecipitation, Immunoaffinity Purification.Specifications
Complement C3 | |
Monoclonal | |
1 mg/ml | |
ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Complement component 3 (C3) purified from human plasma. | |
0.025 mg | |
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
718 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunoprecipitation, Affinity Purification | |
F1-8 | |
Unconjugated | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction