Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589132 Supplier Invitrogen™ Supplier No. PA589132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVTIKPAPET EKRPQDAKNT MILEICTRYR GDQDATMSIL DISMMTGFAP DTDDLKQLAN GVDRYISKYE LDKAFSDRNT LIIYLDKVSH SEDDCLAFKV HQYFNVELIQ PGAVKVYAYY NLEESCTRFY HPEKEDGKLN KLCRDELCRC AEENCFIQKS DDKVTLEERL DKACEPGVDY VYKTRLVKVQ LSNDFDEYIM AIEQTIKSGS DEVQVGQQRT FISPIKCREA LKLEEKKHYL MWGLSSDFWG EKPNLSYIIG KDTWVEHWPE EDECQDEENQ KQCQDLGAFT ESMVVFGCPN; Positive Samples: HepG2, HL-60, HeLa, THP-1, Mouse liver, Rat liver.

C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3
Applications ELISA
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene C3
Gene Accession No. P01024, P01026, P01027
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562)
Gene Symbols C3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12266, 24232, 718
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.