Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3 Recombinant Rabbit Monoclonal Antibody (HL1643)
GREENER_CHOICE

Catalog No. FEMA550326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550326 Supplier Invitrogen™ Supplier No. MA550326
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Complement C3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1643
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene C3
Gene Accession No. P01024, P01026
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562)
Gene Symbols C3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C3. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24232, 718
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.