Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3a/C3a (desArg) Mouse anti-Human, MilliporeSigma™

Catalog No. MABF197825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1978-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1978-25 Supplier MilliporeSigma Supplier No. MABF197825UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Complement C3a/C3a (desArg), clone K13/16 Antibody Neutralizing and Flow Cytometry, for the detection of Complement C3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3a/C3a (desArg)
Applications Immunohistochemistry (Paraffin), Inhibition Assays, Neutralization, Flow Cytometry
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols C3,CPAMD1
Host Species Mouse
Immunogen A full length native C3a fragment purified from human serum that was activated at 37°C f or 1 hour by treatment with 10 mg/mL zymosan.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone K13/16 recognizes complement C3a in human tissues. It targets an epitope that is present on human C3, C3a, and C3a-desArg.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.