Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3a Monoclonal Antibody (2B5)

Catalog No. PIMA547301
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547301 500 μL
PIMA547399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547301 Supplier Invitrogen™ Supplier No. MA547301
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C3a Monoclonal Antibody for Western Blot, ELISA

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3a
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene C3
Gene Accession No. P01024
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3
Gene Symbols C3
Host Species Mouse
Immunogen Recombinant netrin domain of human C3 construct.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.