Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3a Mouse anti-Human, Clone: 600000000, Novus Biologicals™

Mouse Monoclonal Antibody
$274.50 - $1667.00
Specifications
Antigen | Complement C3a |
---|---|
Clone | 600000000 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:1000 - 1:5000, ELISA |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB160022
![]() |
Novus Biologicals
NBP31799850UL |
50 μg |
Each for $274.50
|
|
|||||
NB160020
![]() |
Novus Biologicals
NBP317998100UL |
100 μg |
Each for $400.50
|
|
|||||
NB160021
![]() |
Novus Biologicals
NBP317998500UL |
500 μg |
Each for $1,667.00
|
|
|||||
Description
Complement C3a Monoclonal antibody specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISASpecifications
Complement C3a | |
1 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Neuroscience, Signal Transduction, Stem Cells | |
50% PBS, 50% glycerol | |
718 | |
IgG1 | |
Protein G purified |
600000000 | |
Western Blot 1:1000 - 1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 | |
Recombinant netrin domain of human C3. UniProt: P01024 | |
Primary | |
Store at 4C short term. Store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title