Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3b Monoclonal Antibody (013III-1.16)
GREENER_CHOICE

Catalog No. MA182814
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA182814 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA182814 Supplier Invitrogen™ Supplier No. MA182814
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control for immunohistochemical applications for this product is kidney from post streptococcal glomerulonephritis patients. Mouse anti Human iC3b (Neoantigen) antibody, clone 013III-1.16 recognizes the approximately 42 kDa human inactive complement component 3b (iC3b) neoantigen present in blood serum.

Inactive complement component 3b (iC3b) is a 42kDa neoantigen present in blood serum. Cleavage of C3b into iC3b by the protease factor I, in the presence of cofactors, changes the structure and resulting binding properties, preventing binding by complement factor B and properdin. This prevents further progression of the complement pathway, providing another mechanism by which the complement pathway may be regulated. iC3b is thought to play a role in inducing tolerance, binding complement receptor type 3 on antigen presenting cells and stimulating them to produce transforming growth factor beta2 and interleukin-10. It is thought to stimulate B-cells via the CD21 receptor providing a link between the innate and adaptive immune responses. Additionally iC3b acts as an opsonin which is recognised by complement receptor 3 expressed on leukocytes or complement receptor of the immunoglobulin superfamily (CRIg) on Kupffer cells, stimulating phagocytosis of the tagged cells. iC3b levels are often elevated in diseases such as Systemic Lupus Erythematosis and Rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3b
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 013III-1.16
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation BBS with <0.1% sodium azide
Gene C3
Gene Accession No. P01024
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3a; complement component C3b; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; iC3b; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3
Gene Symbols C3
Host Species Mouse
Immunogen Purified human iC3b.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.