Learn More
Invitrogen™ Complement C3b Monoclonal Antibody (H11)

Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC or -20ºC if preferred. Mouse anti Human C3b antibody, clone H11 recognizes human C3b, part of the complement system.
Specifications
Specifications
| Antigen | Complement C3b |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | H11 |
| Concentration | 50 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | C3 |
| Gene Accession No. | P01024 |
| Gene Alias | Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3a; complement component C3b; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; iC3b; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.