Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Complement C3c Polyclonal Antibody, FITC
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA129718
Description
There is a suggested incubation period of 30-60 minutes at room temperature.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications
Complement C3c | |
Polyclonal | |
FITC | |
C3 | |
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component 3d; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; LOC101108131; LOW QUALITY PROTEIN: complement C3; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
Rabbit | |
IgG fraction | |
RUO | |
100135462, 100860826, 101088220, 101108131, 12266, 24232, 280677, 397072, 443475, 476728, 718 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry | |
0.85 mg/mL | |
PBS with 0.09% sodium azide | |
P01024, P01025, P01026, P01027, P12387, Q2UVX4 | |
C3, LOC101088220, LOC443475 | |
C3c Complement isolated from complement-activated human serum. | |
1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Bovine, Sheep, Pig, Feline, Guinea Pig, Goat, Mink | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction