Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3d Antibody (3), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP151119
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for Immunoprecipitation, Immunoassay.Specifications
Complement C3d | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
3 | |
Unconjugated | |
P01024 | |
C3 | |
The monocolonal antibody 3 (also known as YB2/39-11-1-7) reacts with a linear determinant in C3d. | |
0.05 mg | |
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
718 | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction