Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3d Antibody (C3D/2891), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308764AF594
Description
Complement C3d Monoclonal antibody specifically detects Complement C3d in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Complement C3d | |
Monoclonal | |
Alexa Fluor 594 | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Recombinant fragment (around aa 1250-1450) of human Complement C3d protein (exact sequence is proprietary) (Uniprot: P01024) | |
100 μg | |
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
718 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA, Immunohistochemistry (Paraffin), Protein Array | |
C3D/2891 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction