Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C4a Monoclonal Antibody (24H12)
GREENER_CHOICE

Catalog No. PIMA549440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549440 Supplier Invitrogen™ Supplier No. MA549440
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product was used in an Indirect ELISA.

Complement C4a is a protein that is part of the complement system, which is a group of proteins that work together to help the immune system fight off infections. C4a is produced when the complement system is activated, and it plays a role in inflammation and the recruitment of immune cells to the site of infection. It is also involved in the clearance of immune complexes and the regulation of the complement system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C4a
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 24H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene C4a
Gene Accession No. P0C0L4
Gene Alias C4A; C4AD; CO4A; Complement C4-A
Gene Symbols C4a
Host Species Mouse
Immunogen Recombinant Human C4A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.