Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109468 Supplier Thermo Scientific Supplier No. PA5109468
Add to Cart
Add to Cart

Complement C4 Polyclonal Antibody

Complement C4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C4a
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene C4a
Gene Accession No. P0C0L4
Gene Alias C4A; C4AD; CO4A; Complement C4-A
Gene Symbols C4a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1446 of human Complement C4A (NP_0092242).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.