Learn More
Invitrogen™ Complement C4a Polyclonal Antibody

Description
Immunogen sequence: SCPKEKTTRK KRNVNFQKAI NEKLGQYASP TAKRCCQDGV TRLPMMRSCE QRAARVQQPD CREPFLSCCQ FAESLRKKSR DKGQAGLQRA LEI Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 69%.
Specifications
Specifications
| Antigen | Complement C4a |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C4a |
| Gene Accession No. | P0C0L4 |
| Gene Alias | C4A; C4AD; CO4A; Complement C4-A |
| Gene Symbols | C4a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.