Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C4a Rabbit anti-Human, Clone: 4C3F2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Complement C4a |
---|---|
Clone | 4C3F2 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Complement C4a Monoclonal antibody specifically detects Complement C4a in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Complement C4a | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG | |
Recombinant fusion protein containing a sequence corresponding to amino acids 940-1237 of human Complement C4a (P0C0L4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
4C3F2 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
720 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title