Learn More
Invitrogen™ Complement C4b/C4c Polyclonal Antibody, FITC

Description
PA1-28407 detects C4c / C4b from human samples. Antigen retrieval for IHC (P): Proteinase K 30 min at 37°C.
Specifications
Specifications
| Antigen | Complement C4b/C4c |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide |
| Gene | C4B |
| Gene Accession No. | P0C0L5 |
| Gene Alias | Acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; c4 complement; C4A; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4B (Chido blood group); CPAMD2; CPAMD3; DAMC-258G8.6; MGC164979; RG |
| Gene Symbols | C4B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.