Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C4d Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP231130
Description
Complement C4d Polyclonal specifically detects Complement C4d in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Complement C4d | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1:20-1:80, Immunohistochemistry-Paraffin 1:20-1:80, Immunohistochemistry-Frozen 1:20-1:80 | |
P0C0L4 | |
Rabbit | |
Protein G purified | |
RUO | |
720 | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
PBS (pH 7.4) with 0.1% Proclin 300 | |
C4A | |
Synthetic peptide corresponding to aa1241-1256 of C4 conjugated to KLH | |
0.05 mL | |
Primary | |
Recognizes human complement split product C4d. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction