Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C4d Monoclonal Antibody (LP69), FITC
GREENER_CHOICE

Catalog No. PIMA546581
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546581 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546581 Supplier Invitrogen™ Supplier No. MA546581
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement C4d Monoclonal Antibody for IHC (F)

C4 is a component of the classical complement pathway and is split by activated C1s to C4a and C4b. Together C2a and C4b forms the classical C3 convertase. The molecular mass of C4 is 204 kDa and it consists of 3 subunits: a (97 kDa), b (75 kDa) and g (33 kDa).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C4d
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LP69
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene C4A
Gene Accession No. P0C0L4, P0C0L5
Gene Alias acidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4A; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4D; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); CPAMD2; CPAMD3; DADB-112B14.11; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4
Gene Symbols C4A, C4B
Host Species Mouse
Immunogen Complement C4d from normal human plasma.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100293534, 720, 721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.