Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C5 Antibody (557), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB11012179
Description
Complement C5 Monoclonal specifically detects Complement C5 in Human samples. It is validated for Western Blot, ELISA, Functional, Block/Neutralize.Specifications
Complement C5 | |
Monoclonal | |
0.1 mg/mL | |
Western Blot 1:1000, ELISA 1:100-1:2000, Functional, Immunoassay, Block/Neutralize | |
anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822 | |
Mouse | |
Protein G purified | |
RUO | |
727 | |
Human | |
Purified |
Western Blot, ELISA, Functional Assay, Immunoassay, Neutralization | |
557 | |
Unconjugated | |
P01031 | |
C5 | |
BALB/c mice were immunized with human C5 | |
0.05 mg | |
Primary | |
This reacts with an epitope on C5 and C5a. | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction