Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5a Antibody (C17/5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223498AF532
Description
Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human, Baboon samples. It is validated for Western Blot, ELISA.Specifications
Complement Component C5a | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
C5 | |
Purified human C5a | |
Primary | |
Clone C17/5 reacts with human C5a/C5a(desArg) and cross-reacts with baboon C5a/C5a(desArg). | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
C17/5 | |
Western Blot, ELISA | |
C5, C5A, HE | |
Mouse | |
0.1 mL | |
727 | |
Human, Baboon | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction