Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C5b Monoclonal Antibody (568)
GREENER_CHOICE

Catalog No. 01676536
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-536 Supplier Invitrogen™ Supplier No. MA140163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40163 detects C5/C5b from human samples. MA1-40163 has been successfully used in ELISA and Western blot applications. The MA1-40163 immunogen is an epitope on C5/C5b.

C5b-9 is also known as the TCC. TCC consists of C5b, C6, C7, C8 and C9. The complex forms transmembrane channels which displace lipid molecules and other constituents, thus disrupting the phospholipid bilayer of target cells leading to cell lysis by osmotic leakage. The complex contains neoantigens that are absent from the individual native components and the monoclonal antibody DIA 011-01 is directed against a neoepitope exposed in C9 when incorporated into TCC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C5b
Applications ELISA, Western Blot
Classification Monoclonal
Clone 568
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene C5
Gene Accession No. P01031
Gene Alias anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C5; C5a; C5a anaphylatoxin; C5b; C5D; CHE1; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement C5a anaphylatoxin; complement component 5; complement component C5; CPAMD4; ECLZB; FLJ17816; FLJ17822; MGC142298; prepro-C5
Gene Symbols C5
Host Species Mouse
Immunogen Epitope on C5/C5b.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.