Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C6 Antibody (037), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290102V
Description
Complement C6 Monoclonal specifically detects Complement C6 in Human samples. It is validated for ELISA.Specifications
Complement C6 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
37 | |
ELISA | |
complement component 6, complement component C6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement C6 (AAA59668.1; Met1-Ala934). | |
0.1 mL | |
Immunology, Innate Immunity | |
729 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction