Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C6 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299988AF488
Description
Complement C6 Polyclonal specifically detects Complement C6 in Human samples. It is validated for ELISA.Specifications
Complement C6 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 488 | |
complement component 6, complement component C6 | |
Produced in rabbits immunized with purified, recombinant Complement C6 (AAA59668.1; Met1-Ala934). | |
0.1 mL | |
Immunology, Innate Immunity | |
729 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction