Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C7 Antibody (113), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290244IR
Description
Complement C7 Monoclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISA, ELISASpecifications
Complement C7 | |
Monoclonal | |
DyLight 755 | |
complement component 7, complement component C7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement C7 (P10643; Met1-Gln843). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
113 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
730 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction