Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C7 Antibody (113), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290244MFV500
Description
Complement C7 Monoclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISASpecifications
Complement C7 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
730 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
113 | |
Sandwich ELISA Capture | |
complement component 7, complement component C7 | |
Rabbit immunized with purified, recombinant Human Complement C7 (Uniprot#: P10643, Met1-Gln843) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction