Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C7 Antibody (15D1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280406
Description
Complement C7 Monoclonal specifically detects Complement C7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Complement C7 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200Flow 1:50-1:100 | |
complement component 7, complement component C7 | |
Mouse | |
Protein A purified | |
RUO | |
730 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
15D1 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
C7 | |
Recombinant protein within Human Complement C7 aa 498-641/ 843. (SwissProt: P10643 Human) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction