Learn More
Invitrogen™ Complement C9 Monoclonal Antibody (2A1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA533355
Description
Product is supplied at 1 mL (100 μL/mL). This antibody was shown to compete with antibodies to human C9 for its binding site on the C5b-9 complex, indicating that the reactive epitope is located on the C9 molecule. This product can be used as a coating antibody to detect C5b-9 in plasma and urine samples. Application Notes: ELISA - plates were coated with 30 μg/mL in 50mM carbonatebuffer, pH10.6 for 16h at 4°C. Western Blot - A non-reduced sample treatment and SDS-Page was used. The band size is >200 kDa. IHC-P - Tissue sections fixed in formalin were pretreated with protease type XXIV for 10 minutes at 37°C before incubation. IHC-F- Tissue sections were fixed in acetone for 10 minutes at room temperature before incubation.
C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
Specifications
Complement C9 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
Q62930 | |
C9 | |
Rat C5b-9. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2A1 | |
Unconjugated | |
C9 | |
ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
Mouse | |
Protein G | |
RUO | |
117512 | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.