Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C9 Monoclonal Antibody (C9 53)
GREENER_CHOICE

Catalog No. ABS0045302
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0045302 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0045302 Supplier Thermo Scientific Supplier No. ABS0045302
Add to Cart
Add to Cart

Complement C9 Monoclonal Antibody (C9 53)

Complement C9 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C9
Applications ELISA
Classification Monoclonal
Clone C9 53
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene C9
Gene Accession No. P02748
Gene Alias ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor
Gene Symbols C9
Host Species Mouse
Immunogen Complement component C9.
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 735
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.