Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Complement C9 Monoclonal Antibody (C9-6-25-5)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549001
Description
Complement C9 Monoclonal Antibody for Western Blot
C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
Specifications
Complement C9 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P06683 | |
C9 | |
Murine C9 peptide. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Western Blot | |
C9-6-25-5 | |
Unconjugated | |
C9 | |
ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
Rat | |
Protein G | |
RUO | |
12279 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction