Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qA Antibody (3R9/2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB10064729
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Complement Component C1qA | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:500-1:1000 | |
| C1QA complement component 1, q subcomponent, A chain | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes human complement component 1 q (C1q), a 156kDa secreted protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 3R9/2 | |
| Unconjugated | |
| P02745 | |
| C1QA | |
| Made to Human C1QA | |
| 0.05 mL | |
| Immunology, Innate Immunity | |
| 712 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction