Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qA Antibody (C1QA/2953), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279916F
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Complement Component C1qA | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
C1QA | |
Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) (Uniprot: P02745) | |
0.1 mL | |
Immunology, Innate Immunity | |
Human | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
C1QA/2953 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1QA complement component 1, q subcomponent, A chain | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction