Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qA Antibody (C1QA/2956), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308755UV
Description
Complement Component C1qA Monoclonal antibody specifically detects Complement Component C1qA in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Complement Component C1qA | |
Monoclonal | |
DyLight 350 | |
C1QA complement component 1, q subcomponent, A chain | |
Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) (Uniprot: P02745) | |
100 μg | |
Immunology, Innate Immunity | |
712 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry (Paraffin), Protein Array | |
C1QA/2956 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction