Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qA Antibody (SR1502), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Complement Component C1qA |
---|---|
Clone | SR1502 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Complement Component C1qA Monoclonal antibody specifically detects Complement Component C1qA in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
Complement Component C1qA | |
Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
C1QA complement component 1, q subcomponent, A chain | |
Complement Component C1qA (Uniprot #: P02745) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1502 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
712 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title