Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qA, Mouse anti-Human, Clone: C1QA/2783, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Complement Component C1qA |
---|---|
Clone | C1QA/2783 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, ELISA, Protein Array |
Applications | Western Blot, ELISA, Peptide Array |
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
Complement Component C1qA | |
0.2 mg/mL | |
Western Blot, ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity | |
10 mM PBS with 0.05% BSA | |
C1QA complement component 1, q subcomponent, A chain | |
C1QA | |
IgG2b κ | |
Protein A or G purified |
C1QA/2783 | |
Western Blot 1-2 ug/ml, ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P02745, P02745 | |
712 | |
Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title