Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1qC Antibody (4H9D7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
Antigen | Complement Component C1qC |
---|---|
Clone | 4H9D7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61867
![]() |
Novus Biologicals
NBP261867 |
0.1 mL |
Each of 1 for $562.50
|
|
|||||
Description
Complement Component C1qC Monoclonal specifically detects Complement Component C1qC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Complement Component C1qC | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
714 | |
Purified recombinant fragment of human C1QC (AA: 115-245) expressed in E. Coli. | |
Primary | |
25.7 kDa |
4H9D7 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
C1Q-C, C1QGcomplement component 1, q subcomponent, gamma polypeptide, complement C1q subcomponent subunit C, complement component 1, q subcomponent, C chain, FLJ27103 | |
C1QC | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title