Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C1r Antibody (OTI1F1), DyLight 650, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270318C

Catalog No. NBP270318C


Only null left
Add to Cart

Description

Description

Complement Component C1r Monoclonal specifically detects Complement Component C1r in Human, Canine samples. It is validated for Western Blot.
Specifications

Specifications

Complement Component C1r
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
C1R
Recombinant protein fragment corresponding to amino acids 262-562 of human Complement Component C1r (NP_001724) produced in E.coli.
0.1 mL
715.0
Store at 4°C in the dark.
Western Blot
OTI1F1
Western Blot
complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41
Mouse
Affinity Purified
Primary
Human, Canine
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.