Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1r Antibody (OTI1F1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP203821
Description
Complement Component C1r Monoclonal specifically detects Complement Component C1r in Human, Canine samples. It is validated for Western Blot.Specifications
Complement Component C1r | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-2000 | |
C1R | |
Human recombinant protein fragment corresponding to amino acids 262-562 of human C1R (NP_001724) produced in E.coli. | |
Affinity Purified | |
RUO | |
715 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
OTI1F1 | |
Unconjugated | |
complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41 | |
Mouse | |
80 kDa | |
0.1 mL | |
Primary | |
Human, Canine | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction