Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1r Antibody (OTI1F1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270318PE
Description
Complement Component C1r Monoclonal antibody specifically detects Complement Component C1r in Human, Canine samples. It is validated for Western BlotSpecifications
Complement Component C1r | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
715 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot | |
OTI1F1 | |
Western Blot | |
complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41 | |
Human recombinant protein fragment corresponding to amino acids 262-562 of human C1R (NP_001724) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction